CAS RN 18246-28-1
IUPAC Name:
(5-bromothiophen-2-yl)trimethylsilane
Synonyms:
(5-bromothiophen-2-yl)trimethylsilane
Molecular Weight (g/mol):
235.21
Molecular Formula:
C7H11BrSSi
InChi Key:
IXLYRDOGOXBECE-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C1=CC=C(Br)S1