accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHOH2NOO(E/Z)

CAS RN 1824819-33-1

IUPAC Name: (9H-fluoren-9-yl)methyl 4-(N'-hydroxycarbamimidoyl)piperidine-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 4-(N'-hydroxycarbamimidoyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 365.43
Molecular Formula: C21H23N3O3
InChi Key: ROHLBMBCFADFMX-UHFFFAOYSA-N
SMILES: NC(=NO)C1CCN(CC1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results