accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 182500-26-1

IUPAC Name: 1-isocyanato-2-(trifluoromethoxy)benzene
Synonyms: 1-isocyanato-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 203.12
Molecular Formula: C8H4F3NO2
InChi Key: CTMGYQHKKIEXKF-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1N=C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results