accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18253-54-8

IUPAC Name: 1,1-dichloro-2,11,20,29,38,39,40,41-octaaza-1-stannadodecacyclo[19.17.1.1¹,¹².1³,³⁷.0²,¹⁰.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0²⁸,³⁹.0³⁰,³⁸.0³¹,³⁶.0¹⁹,⁴¹]hentetraconta-3,5,7,9,11,13,15,17,19,21,23,25,27,29,31,33,35,37(40)-octadecaene
Synonyms: 1,1-dichloro-2,11,20,29,38,39,40,41-octaaza-1-stannadodecacyclo[19.17.1.1¹,¹².1³,³⁷.0²,¹⁰.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0²⁸,³⁹.0³⁰,³⁸.0³¹,³⁶.0¹⁹,⁴¹]hentetraconta-3,5,7,9,11,13,15,17,19,21,23,25,27,29,31,33,35,37(40)-octadecaene
Molecular Weight (g/mol): 702.15
Molecular Formula: C32H16Cl2N8Sn
InChi Key: SDIGTTATIKXMSE-UHFFFAOYSA-L
SMILES: Cl[Sn]123(Cl)N4C5=NC6=C7C=CC=CC7=C(N=C7N1C(=NC1=C8C=CC=CC8=C(N=C4C4=CC=CC=C54)N21)C1=CC=CC=C71)N36
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results