accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1826-11-5

IUPAC Name: 2-phenyl-1,3-thiazole
Synonyms: thiazole, 2-phenyl-
Molecular Weight (g/mol): 161.22
Molecular Formula: C9H7NS
InChi Key: WYKHSBAVLOPISI-UHFFFAOYSA-N
SMILES: S1C=CN=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results