accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OZr

CAS RN 18267-08-8

IUPAC Name: zirconium(4+) ethanolate
Synonyms: zirconium(4+) ethoxide
Molecular Weight (g/mol): 136.28
Molecular Formula: C2H5OZr
InChi Key: VCYRFVPNTBNCLW-UHFFFAOYSA-N
SMILES: [Zr+4].CC[O-]
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)