accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2F

CAS RN 1827-27-6

IUPAC Name: 6-fluoropyridin-3-amine
Synonyms: 5-amino-2-fluoropyridine
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H5FN2
InChi Key: YTHMOBMZVVFNBE-UHFFFAOYSA-N
SMILES: NC1=CN=C(F)C=C1
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)