accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18270-61-6

IUPAC Name: 2-[chloro(phenyl)methylidene]propanedinitrile
Synonyms: 2-[chloro(phenyl)methylidene]propanedinitrile
Molecular Weight (g/mol): 188.61
Molecular Formula: C10H5ClN2
InChi Key: HGWXUOJXKLGEAF-UHFFFAOYSA-N
SMILES: ClC(=C(C#N)C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results