accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 18281-04-4

IUPAC Name: ethyl nonadecanoate
Synonyms: ethyl nonadecanoate
Molecular Weight (g/mol): 326.57
Molecular Formula: C21H42O2
InChi Key: ICVYQLQYVCXJNE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC(=O)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results