accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1829-00-1

IUPAC Name: disodium 6-methyl-2-(4-{3-[4-(6-methyl-7-sulfonato-1,3-benzothiazol-2-yl)phenyl]triaz-2-en-1-yl}phenyl)-1,3-benzothiazole-7-sulfonate
Synonyms: titan yellow
Molecular Weight (g/mol): 695.71
Molecular Formula: C28H19N5Na2O6S4
InChi Key: CZIRZNRQHFVCDZ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CC1=CC=C2N=C(SC2=C1S([O-])(=O)=O)C1=CC=C(NN=NC2=CC=C(C=C2)C2=NC3=CC=C(C)C(=C3S2)S([O-])(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results