accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 182918-13-4

IUPAC Name: 2,4-bis({[1,1'-biphenyl]-4-yl})-6-chloro-1,3,5-triazine
Synonyms: 2,4-bis({[1,1'-biphenyl]-4-yl})-6-chloro-1,3,5-triazine
Molecular Weight (g/mol): 419.91
Molecular Formula: C27H18ClN3
InChi Key: FVQBRDRAILXTMJ-UHFFFAOYSA-N
SMILES: ClC1=NC(=NC(=N1)C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results