accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOCH3

CAS RN 1830-54-2

IUPAC Name: 1,5-dimethyl 3-oxopentanedioate
Synonyms: 1,5-dimethyl 3-oxopentanedioate
Molecular Weight (g/mol): 174.15
Molecular Formula: C7H10O5
InChi Key: RNJOKCPFLQMDEC-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)CC(=O)OC
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)