accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOOHOOCH3CH2

CAS RN 1830-78-0

IUPAC Name: 2-hydroxy-3-[(2-methylprop-2-enoyl)oxy]propyl 2-methylprop-2-enoate
Synonyms: glyceryl dimethacrylate
Molecular Weight (g/mol): 228.24
Molecular Formula: C11H16O5
InChi Key: OQHMGFSAURFQAF-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(O)COC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results