CAS RN 183072-30-2
CAS RN 183072-30-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich (2R)-2-Phenyl-3,4-Dihydro-2H-Pyrimido[2,1-B][1,3]Benzothiazole
Sigma Aldrich (2R)-2-Phenyl-3,4-Dihydro-2H-Pyrimido[2,1-B][1,3]Benzothiazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.