CAS RN 1831130-33-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%
2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%
CAS: 1831130-33-6 Synonym: 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,2-amino-3-fluorophenylboronic acid pinacol ester,2-amino-3-fluorobenzeneboronic acid pinacol ester,2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine