accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 18312-04-4

IUPAC Name: octanoic acid
Synonyms: caprylic acid zinc salt caprylic acid lanthanum(+3) salt caprylic acid caprylic acid, 14c-labeled lead(+2) salt caprylic acid cobalt salt caprylic acid kortacid 0899 lithium salt caprylic acid aliphatic carboxylic acid, c8:0 acide octanoique
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: WWZKQHOCKIZLMA-UHFFFAOYSA-N
SMILES: CCCCCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results