accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 183266-61-7

IUPAC Name: 1-(1H-1,2,3-benzotriazol-1-yl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(1,2,3-benzotriazol-1-yl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 215.14
Molecular Formula: C8H4F3N3O
InChi Key: GVQIQOIKWUOEJP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)N1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results