accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2COSiCH3CH3H3C

CAS RN 1833-53-0

IUPAC Name: trimethyl(prop-1-en-2-yloxy)silane
Synonyms: trimethyl(prop-1-en-2-yloxy)silane
Molecular Weight (g/mol): 130.26
Molecular Formula: C6H14OSi
InChi Key: UAIFZYSPVVBOPN-UHFFFAOYSA-N
SMILES: CC(=C)O[Si](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results