accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 183582-92-5

IUPAC Name: cyclohexane-1,3,5-tricarbonitrile
Synonyms: cyclohexane-1,3,5-tricarbonitrile
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: VMUOSHREZKXCIV-UHFFFAOYSA-N
SMILES: N#CC1CC(CC(C1)C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results