CAS RN 1836-06-2
IUPAC Name:
1-(3-bromo-4-hydroxyphenyl)ethan-1-one
Synonyms:
1-(3-bromo-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol):
215.05
Molecular Formula:
C8H7BrO2
InChi Key:
LEXAYGVHWYOOAB-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(Br)=C(O)C=C1
1
–
1
of
1
results
1
3'-Bromo-4'-hydroxyacetophenone 98.0+%, TCI America™
CAS: 1836-06-2 Molecular Formula: C8H7BrO2 Molecular Weight (g/mol): 215.046 MDL Number: MFCD05663950 InChI Key: LEXAYGVHWYOOAB-UHFFFAOYSA-N PubChem CID: 608094 IUPAC Name: 1-(3-bromo-4-hydroxyphenyl)ethanone SMILES: CC(=O)C1=CC(=C(C=C1)O)Br