accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18362-51-1

IUPAC Name: 1,3-bis(4-methoxyphenyl)propane-1,3-dione
Synonyms: 1,3-bis(4-methoxyphenyl)propane-1,3-dione
Molecular Weight (g/mol): 284.31
Molecular Formula: C17H16O4
InChi Key: GNMDORSUZRRMFS-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results