accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONOOHNH2

CAS RN 183673-71-4

IUPAC Name: 4-amino-1-[(tert-butoxy)carbonyl]piperidine-4-carboxylic acid
Synonyms: 4-amino-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid
Molecular Weight (g/mol): 244.29
Molecular Formula: C11H20N2O4
InChi Key: YNHLVALLAURVJF-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results