accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HONb

CAS RN 18368-80-4

IUPAC Name: pentakis(propan-2-ol) niobium
Synonyms: pentakis(isopropyl alcohol) niobium
Molecular Weight (g/mol): 393.39
Molecular Formula: C15H40NbO5
InChi Key: FTYCGXPVPAOYKN-UHFFFAOYSA-N
SMILES: [Nb].CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results