accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1837-91-8

IUPAC Name: 1,2,3,4,5,6-hexabromocyclohexane
Synonyms: 1,2,3,4,5,6-hexabromocyclohexane
Molecular Weight (g/mol): 557.54
Molecular Formula: C6H6Br6
InChi Key: QFQZKISCBJKVHI-UHFFFAOYSA-N
SMILES: BrC1C(Br)C(Br)C(Br)C(Br)C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results