CAS RN 18372-22-0
IUPAC Name:
methyl 2-(1H-indol-3-yl)-2-oxoacetate
Synonyms:
methyl 2-(1H-indol-3-yl)-2-oxoacetate
Molecular Weight (g/mol):
203.2
Molecular Formula:
C11H9NO3
InChi Key:
VFIJGAWYVXDYLK-UHFFFAOYSA-N
SMILES:
COC(=O)C(=O)C1=CNC2=CC=CC=C12
1
–
1
of
1
results
1
Sigma Aldrich Methyl 3-indoleglyoxylate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.