accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1838-08-0

IUPAC Name: octadecan-1-aminium chloride
Synonyms: octadecan-1-aminium chloride
Molecular Weight (g/mol): 305.98
Molecular Formula: C18H40ClN
InChi Key: RNYJXPUAFDFIQJ-UHFFFAOYSA-N
SMILES: [Cl-].CCCCCCCCCCCCCCCCCC[NH3+]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results