accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1838-41-1

IUPAC Name: dichloroiodanuide; tetramethylazanium
Synonyms: dichloroiodanuide; tetramethylammonium ion
Molecular Weight (g/mol): 271.95
Molecular Formula: C4H12Cl2IN
InChi Key: FUNMSHCNGVSXHD-UHFFFAOYSA-N
SMILES: Cl[I-]Cl.C[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results