accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OHONHCH3OH3CCH3H3CO

CAS RN 183990-64-9

IUPAC Name: (3R)-3-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid
Synonyms: (3R)-3-[(tert-butoxycarbonyl)amino]-4-methylpentanoic acid
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: LUXMZCJCTUATDM-MRVPVSSYSA-N
SMILES: CC(C)[C@@H](CC(O)=O)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)