accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1840-19-3

IUPAC Name: 1-isothiocyanato-3-(trifluoromethyl)benzene
Synonyms: 1-isothiocyanato-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 203.18
Molecular Formula: C8H4F3NS
InChi Key: GFEPANUKFYVALF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)N=C=S
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)