accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18401-43-9

IUPAC Name: {bicyclo[2.2.1]hept-5-en-2-yl}triethoxysilane
Synonyms: bicyclo[2.2.1]hept-5-en-2-yltriethoxysilane
Molecular Weight (g/mol): 256.42
Molecular Formula: C13H24O3Si
InChi Key: HHKDWDAAEFGBAC-UHFFFAOYNA-N
SMILES: CCO[Si](OCC)(OCC)C1CC2CC1C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results