accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 184026-06-0

IUPAC Name: 6-nitronaphthalene-2,3-dicarbonitrile
Synonyms: 6-nitronaphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol): 223.19
Molecular Formula: C12H5N3O2
InChi Key: JJSGXASWKOLCMQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C=C(C#N)C(=CC2=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results