accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiSiOH3COH3COH3COH3COH3C

CAS RN 18406-41-2

IUPAC Name: 3,3,6,6-tetramethoxy-2,7-dioxa-3,6-disilaoctane
Synonyms: hexamethoxydisilylethane
Molecular Weight (g/mol): 270.43
Molecular Formula: C8H22O6Si2
InChi Key: JCGDCINCKDQXDX-UHFFFAOYSA-N
SMILES: CO[Si](CC[Si](OC)(OC)OC)(OC)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results