accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18418-08-1

IUPAC Name: ethyl 2-hydroxybut-3-ynoate
Synonyms: ethyl 2-hydroxybut-3-ynoate
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: GDWLVFKOPXSIAE-UHFFFAOYNA-N
SMILES: CCOC(=O)C(O)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results