accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1844-09-3

IUPAC Name: 1-(octadecylsulfanyl)octadecane
Synonyms: 1-(octadecylsulfanyl)octadecane
Molecular Weight (g/mol): 539.05
Molecular Formula: C36H74S
InChi Key: IHWDIGHWDQPQMQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCSCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results