accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1844839-22-0

IUPAC Name: 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Synonyms: 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular Weight (g/mol): 250.08
Molecular Formula: C13H16BFO3
InChi Key: DGVLPOACSLDJBC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(C=O)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results