accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1845-25-6

IUPAC Name: 2-hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one
Synonyms: oxypinocamphone
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: VZRRCQOUNSHSGB-UHFFFAOYNA-N
SMILES: CC1(C)C2CC1C(C)(O)C(=O)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results