accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OO

CAS RN 18456-87-6

IUPAC Name: 5-(propan-2-yl)cyclohexane-1,3-dione
Synonyms: 5-isopropylcyclohexane-1,3-dione
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: DUKZQEYDAFUJMK-UHFFFAOYSA-N
SMILES: CC(C)C1CC(=O)CC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results