accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3COOOOSiCH3CH3H3C

CAS RN 18457-04-0

IUPAC Name: 1,3-ditrimethylsilyl propanedioate
Synonyms: 1,3-ditrimethylsilyl propanedioate
Molecular Weight (g/mol): 248.43
Molecular Formula: C9H20O4Si2
InChi Key: ATCKJLDGNXGLAO-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=O)CC(=O)O[Si](C)(C)C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)