accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOONH2

CAS RN 184698-41-7

IUPAC Name: methyl 4-aminooxane-4-carboxylate
Synonyms: methyl 4-aminooxane-4-carboxylate
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: AZFBPYNPITWILD-UHFFFAOYSA-N
SMILES: COC(=O)C1(N)CCOCC1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results