accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18479-51-1

IUPAC Name: 3,7-dimethyloct-6-en-3-ol
Synonyms: 1,2-dihydrolinalool
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: JRTBBCBDKSRRCY-UHFFFAOYNA-N
SMILES: CCC(C)(O)CCC=C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results