accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO2H2H2H

CAS RN 1849-29-2

IUPAC Name: (²H₃)methanol
Synonyms: (²H₃)methanol
Molecular Weight (g/mol): 35.06
Molecular Formula: CH4O
InChi Key: OKKJLVBELUTLKV-FIBGUPNXSA-N
SMILES: [2H]C([2H])([2H])O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results