accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 18495-82-4

IUPAC Name: 4-(3-phenylpropyl)piperidine
Synonyms: 4-(3-phenylpropyl)piperidine
Molecular Weight (g/mol): 203.33
Molecular Formula: C14H21N
InChi Key: HASRFXGIJALRRB-UHFFFAOYSA-N
SMILES: C(CC1CCNCC1)CC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results