accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18503-90-7

IUPAC Name: (dibutoxymethyl)dimethylamine
Synonyms: (dibutoxymethyl)dimethylamine
Molecular Weight (g/mol): 203.33
Molecular Formula: C11H25NO2
InChi Key: GSFFXKGTGPMVLM-UHFFFAOYSA-N
SMILES: CCCCOC(OCCCC)N(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results