accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 185041-05-8

IUPAC Name: methyl 2-chloro-4-iodopyridine-3-carboxylate
Synonyms: methyl 2-chloro-4-iodopyridine-3-carboxylate
Molecular Weight (g/mol): 297.48
Molecular Formula: C7H5ClINO2
InChi Key: FXGPTKSMXUWJGO-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)N=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results