accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONOSOOFFF

CAS RN 185099-68-7

IUPAC Name: tert-butyl 3-(trifluoromethanesulfonyloxy)-8-azabicyclo[3.2.1]oct-2-ene-8-carboxylate
Synonyms: tert-butyl 3-(trifluoromethanesulfonyloxy)-8-azabicyclo[3.2.1]oct-2-ene-8-carboxylate
Molecular Weight (g/mol): 357.34
Molecular Formula: C13H18F3NO5S
InChi Key: CJDKVEWDWXIBSM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1C2CCC1C=C(C2)OS(=O)(=O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)