CAS RN 185112-61-2
IUPAC Name:
9-(3-bromophenyl)-9H-carbazole
Synonyms:
9-(3-bromophenyl)carbazole
Molecular Weight (g/mol):
322.21
Molecular Formula:
C18H12BrN
InChi Key:
ZKGHGKNHPPZALY-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
9-(3-Bromophenyl)carbazole 98.0+%, TCI America™
CAS: 185112-61-2 Molecular Formula: C18H12BrN Molecular Weight (g/mol): 322.205 MDL Number: MFCD20486475 InChI Key: ZKGHGKNHPPZALY-UHFFFAOYSA-N PubChem CID: 58690888 IUPAC Name: 9-(3-bromophenyl)carbazole SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC(=CC=C4)Br