CAS RN 185116-42-1
IUPAC Name:
3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)benzoic acid
Synonyms:
3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}benzoic acid
Molecular Weight (g/mol):
359.38
Molecular Formula:
C22H17NO4
InChi Key:
VFXDSSUGFNVDJP-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)=C1
1
–
1
of
1
results
1
Sigma Aldrich 6-Ethoxy-6-Oxohexylzinc Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.