accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHNOO

CAS RN 185116-43-2

IUPAC Name: 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)benzoic acid
Synonyms: 4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}benzoic acid
Molecular Weight (g/mol): 359.38
Molecular Formula: C22H17NO4
InChi Key: VGSYYBSAOANSLR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)