CAS RN 185137-29-5
IUPAC Name:
4-phenyl-1,3-thiazolidine-2-thione
Synonyms:
4-phenyl-1,3-thiazolidine-2-thione
Molecular Weight (g/mol):
195.3
Molecular Formula:
C9H9NS2
InChi Key:
IEXSISKCCADMLK-UHFFFAOYNA-N
SMILES:
S=C1NC(CS1)C1=CC=CC=C1
1
–
1
of
1
results
1
(S)-4-Phenylthiazolidine-2-thione 98.0+%, TCI America™
CAS: 185137-29-5 Molecular Formula: C9H9NS2 Molecular Weight (g/mol): 195.298 MDL Number: MFCD06658214 InChI Key: IEXSISKCCADMLK-MRVPVSSYSA-N PubChem CID: 11333042 IUPAC Name: (4S)-4-phenyl-1,3-thiazolidine-2-thione SMILES: C1C(NC(=S)S1)C2=CC=CC=C2