accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 185147-08-4

IUPAC Name: 4-fluoro-3-methylbenzonitrile
Synonyms: 4-fluoro-3-methylbenzonitrile
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: ZMEAHKIIWJDJFT-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1F)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results